Learn More
Lutetium(III) chloride hexahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
108.00 € - 338.00 €
Chemical Identifiers
CAS | 15230-79-2 |
---|---|
Molecular Formula | H3LuN3O9 |
Molecular Weight (g/mol) | 364.00 |
MDL Number | MFCD00149775 |
InChI Key | MESQZVHAWZZUAY-UHFFFAOYSA-N |
IUPAC Name | lutetium(3+) tris(nitric acid) |
SMILES | [Lu+3].O[N+]([O-])=O.O[N+]([O-])=O.O[N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11336328
|
Thermo Scientific Alfa Aesar
011260.03 |
1 g |
108.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11326328
|
Thermo Scientific Alfa Aesar
011260.06 |
5 g |
338.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lutetium(III) chloride hexahydrate is used as a catalyst and laboratory reagent. It is also used in ceramics, optical glasses, electrical components and photo-optical material. It is one of the components of scintillating composition. It serves as a precursor for the preparation of other lutetium complexes. It finds application as one of the starting materials for aluminate phosphors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15230-79-2 | |
364.00 | |
MESQZVHAWZZUAY-UHFFFAOYSA-N | |
[Lu+3].O[N+]([O-])=O.O[N+]([O-])=O.O[N+]([O-])=O |
H3LuN3O9 | |
MFCD00149775 | |
lutetium(3+) tris(nitric acid) |
Specifications
15230-79-2 | |
H3LuN3O9 | |
14,5615 | |
MESQZVHAWZZUAY-UHFFFAOYSA-N | |
lutetium(3+) tris(nitric acid) | |
389.42 (281.33 Anhydrous) | |
Reacton™ | |
Lutetium(III) chloride hexahydrate |
Crystalline Aggregates | |
MFCD00149775 | |
Soluble in water. | |
[Lu+3].O[N+]([O-])=O.O[N+]([O-])=O.O[N+]([O-])=O | |
364.00 | |
99.9% | |
(REO) |
Safety and Handling
missing translation for 'einecsNumber' : 233-240-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only