missing translation for 'onlineSavingsMsg'
Learn More

Lutetium(III) fluoride, anhydrous, 99.9% (REO), Thermo Scientific™

For vacuum deposition

Chemical Identifiers

CAS 13760-81-1
MDL Number MFCD00016191
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11364118
View Documents Promotion Details
Thermo Scientific Alfa Aesar
041415.03
1 g N/A
N/A
11374118
View Documents Promotion Details
Thermo Scientific Alfa Aesar
041415.06
5 g N/A
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

13760-81-1
MFCD00016191

Specifications

1182°C
Pieces
UN3288
99.9%
Hygroscopic
(REO)
13760-81-1
MFCD00016191
231.97
Odorless
8.3 g/mL
Lutetium(III) fluoride, Anhydrous
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 237-355-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications