missing translation for 'onlineSavingsMsg'
Learn More

Lutetium(III) fluoride, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11372578
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13760-81-1
MDL Number MFCD00016191

Specifications

Melting Point 1182°C
Physical Form Lump
Quantity 1 g
UN Number UN3288
Formula Weight 231.97
Percent Purity 99.9%
Grade Reacton™
Odor Odorless
Sensitivity Hygroscopic
Density 8.3 g/mL
Assay (REO)
Chemical Name or Material Lutetium(III) fluoride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'