missing translation for 'onlineSavingsMsg'
Learn More

Lutetium(III) oxide, 99.9% (REO), Thermo Scientific™

Product Code. p-7050231
Click to view available options
Quantity:
1 g
2 g
5 g
Unit Size:
1g
2g
5g
This item is not returnable. View return policy

Product Code. 11313308

Brand: Thermo Scientific Alfa Aesar 036244.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12032-20-1
Molecular Formula Lu2O3
Molecular Weight (g/mol) 397.93
MDL Number MFCD00011100
InChI Key UGBIHFMRUDAMBY-UHFFFAOYSA-N
Synonym lutetium 3+ ; oxygen 2-, lutetium oxide, lutetium iii oxide trace rare earth metals basis 1g, cid159406, einecs 234-764-3, lu2o3, lutetium 3+
IUPAC Name dilutetium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Lu+3].[Lu+3]

Specifications

Melting Point 2487°C
Physical Form Sintered Lump
Quantity 1 g
Merck Index 14,5615
Solubility Information Insoluble in water.
Formula Weight 397.94
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 9.420 g/mL
Assay (REO)
Chemical Name or Material Lutetium(III) oxide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.