Learn More
Invitrogen™ Lyo-ready Enzymes and Reagents for Recombinase Polymerase Amplification (RPA)
Invitrogen Lyo-ready RPA reagents are designed for fast, sensitive, and specific DNA amplification through isothermal recombinase polymerase amplification (RPA) reactions. Lyo-ready RPA reagents are available in stand-alone and kit formats.
76.70 € - 1070.00 €
Specifications
Form | Liquid |
---|---|
Research Category | Nucleic Acid Amplification |
Shipping Condition | Dry Ice |
Product Code | Brand | Product Name | Quantity | Product Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Name | Quantity | Product Type | Price | Quantity & Availability | |||
30245335
|
Invitrogen™
A72123 |
Lyo-ready T4 Gene 32 Protein | 1600 μg | Stand-alone enzyme |
1070.00 €
1600µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
30245214
|
Invitrogen™
A72124 |
Lyo-ready T4 UvsX Protein | 120 μg | Stand-alone enzyme |
173.00 €
120µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
30245152
|
Invitrogen™
A72125 |
Lyo-ready T4 UvsY Protein | 120 μg | Stand-alone enzyme |
210.00 €
120µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
30245027
|
Invitrogen™
A72126 |
RPA Reaction Buffer (2X) | 2000 μL | Reaction Buffer |
76.70 €
2000µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
30245142
|
Invitrogen™
A72127 |
Lyo-ready RPA Kit | 100 Reactions | DNA Amplification Kit |
246.00 €
100 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Invitrogen Lyo-ready recombinase polymerase amplification (RPA) reagents are designed for fast, sensitive, and specific DNA amplification through isothermal RPA reactions. Lyo-ready RPA reagents are available in stand-alone and kit formats. Reverse transcriptase can be added to the RPA reaction for amplification of RNA targets. All reagents are glycerol-free, which enables assay lyophilization.
Features of Lyo-ready RPA products
- Enhanced sensitivity down to 1 copy using optimized (RT-)RPA reaction buffer
- Robust and specific RNA/DNA amplification
- Amplification from inhibitor-containing samples
- Fast target amplification in as little as 20 minutes
- Ability to easily optimize RPA or RT-RPA reactions
Kit components
The Lyo-ready RPA Kit includes all required components for an RPA reaction:
- Lyo-ready T4 UvsX Protein—produced from E. coli cells bearing cloned uvsX gene derived from bacteriophage T4
- Lyo-ready T4 UvsY Protein—a recombination mediator protein that stimulates homologous recombination performed by the UvsX recombinase
- Lyo-ready Bst DNA Polymerase (Cat. No. A56655)—a proprietary mutant of Bst DNA Polymerase, Large Fragment, with fast reaction speed, increased sensitivity, and tolerance to inhibitors
- Lyo-ready T4 Gene 32 Protein—a single-stranded DNA binding protein produced from E. coli cells carrying cloned bacteriophage T4 gene 32
- Optimized reaction buffer, magnesium chloride, and dNTPs
Specifications
Liquid | |
Nucleic Acid Amplification | |
Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.