missing translation for 'onlineSavingsMsg'
Learn More

LYPD5 293T Cell Overexpression Lysate (Denatured), Abnova

Brand:  Abnova H00284348-T01.100uL


Product Code. 16179943

  • 215.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Western Blotting

Specifications

Specifications

293T
LYPD5 293T Cell Transient Overexpression Lysate(Denatured)
27.5
pCMV-LYPD5 full-length
100 μL
Human
Western Blot
BC057816
6.8
Transient overexpression cell Lysate was tested with Anti-LYPD5 antibody (H00284348-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 100UL LYPD5 293T Cell Transient Overexpression Lys

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

Supplemental information
  • MIXTURE LIST-Contains : Mercaptoethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers