Learn More
Labconco™ Lyph-Lock™ Freeze-Dry Flask Assembly
Complete three-piece flask consists of high-strength borosilicate glass molded top and bottom with silicone rubber seal
Brand:  Labconco™ 7554900
 View more versions of this product
Product Code. 10053612
  • 595.00 € / Each
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Complete three-piece flask consists of high-strength borosilicate glass molded top and bottom with silicone rubber seal
  • With 24/40 joints
  • Seal grasps top and bottom securely—never comes in contact with sample
  • Flat bottomed
  • Wide mouth lets you load samples easily and lyophilize quickly
  • Compatible with Labconco, Virtis™ and other major brands of freeze dryers
  • Flasks should be filled to no more than one-third the volume so that maximum surface area is achieved for efficient lyophilization
Required Accessories
  • Adapters are required to attach flasks to freeze dry valve ports
Specifications
Specifications
3.55 in.
Glass
14.5 cm
24/40
90.2 mm
750 mL
5.7 in.
Complete three-piece flask consists of high-strength Borosilicate Glass molded top and bottom with Silicone Rubber seal
3 lb.
1.4 kg