Learn More
Invitrogen™ Lysing Solution, Whole Blood Lysing Solution
Lysing Solution, Whole Blood Lysing Solution
Brand:  Invitrogen™ GAS010S100
 View more versions of this product
Product Code. 10292873
  • 734.00 € / Each
Explore more special offers
Description
Description
Invitrogen Cal-Lyse™ is a lysing solution formulated to enable the lysis of erythrocytes in samples of anticoagulated human peripheral blood. Invitrogen Cal-Lyse™ lysing solution is intended to be used as an aid in the enumeration of leukocytes that have been stained with monoclonal antibodies for flow cytometric analysis.
View a selection guide for all cell lysis and preservation solutions and products related to sample fixation and permeablization, blocking reagents, and information about the use of Dynabeads™ for cell isolation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store at room temperature.
  • Shelf life: 24 months from date of manufacture
  • Cell Lysis Buffer
    100 mL
    100 mL
    Room Temperature
    Whole Blood Lysing Solution