Learn More
Melford Lysozyme from Chicken Egg White
Purified from chicken egg white, 2x crystallized, dialyzed and supplied as a salt free lyophilized powder.
90.85 € - 426.00 €
Specifications
CAS 12650-88-3
View More Specs
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30018447
View Documents
Melford
L38100-1.0
1 g
90.85 €
1g
30018449
View Documents
Melford
L38100-5.0
5 g
286.00 €
5g
30018448
View Documents
Melford
L38100-10.0
10 g
426.00 €
10g
Description
Description
Purified from chicken egg white, 2x crystallized, dialyzed and supplied as a salt free lyophilized powder.
Purified from chicken egg white, 2x crystallized, dialyzed and supplied as a salt free lyophilized powder.
Dry powder stable for 3 - 5 years when stored dry at 2°C to 8°C.
Stock solution is stable up to 3 days at 4°C and up to 1 year at -20°C.
Specifications
Specifications
12650-88-3