missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals m-Cresol Purple, pure, indicator grade

Product Code. p-94183 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
5g
This item is not returnable. View return policy

Product Code. 10792461

Brand: Thermo Scientific Chemicals 228200050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2303-01-7 | C21H18O5S | 382.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2303-01-7
Molecular Formula C21H18O5S
Molecular Weight (g/mol) 382.43
MDL Number MFCD00005871
InChI Key OLQIKGSZDTXODA-UHFFFAOYSA-N
PubChem CID 73030
IUPAC Name 4-[3-(4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
SMILES CC1=C(C=CC(=C1)O)C2(C3=CC=CC=C3S(=O)(=O)O2)C4=C(C=C(C=C4)O)C

Specifications

Infrared Spectrum Authentic
Formula Weight 382.42
Grade Pure
Packaging Glass bottle
Specific Extinction 900 min. (578nm, in buffer)
Visual Transition Interval from pH 7.4 (yellow) to pH 9.0 (purple)
Color Green
Quantity 5 g
Physical Form Fine Crystalline Powder
Chemical Name or Material m-Cresol Purple

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.