missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Branson Ultrasonics™ M Series Ultrasonic Cleaning Bath
Outfit your lab with a Bransonic MH Series Ultrasonic Cleaner. This unit is designed with solid construction and excellent functionality for a variety of applications − including cell separation, sample preparation and degassing of liquids.
Brand: Branson Ultrasonics™ CPX-952-536R
This item is not returnable.
View return policy
Description
Outfit your lab with a Bransonic MH Series Ultrasonic Cleaner. This unit is designed with solid construction and excellent functionality for a variety of applications − including cell separation, sample preparation and degassing of liquids.
- Sweep frequency eliminates standing waves and creates consistent cavitation throughout the tank, eliminating dead zones for long life
- Includes 60-minute mechanical timer and continuous operation
- 40kHz industrial transducers
- Controls located above and behind in easily accessible control tower panel which is raised above tank to avoid damage from cleaning solutions
- Plastic body
- Chemical-resistant
- 1.5 gal. and larger models
- Tank drains with valves
Specifications
None - Control Dial | |
60 min. | |
230/240 V | |
9.5 L | |
Tank cover | |
14 lb. | |
2 Years |
Plastic Housing, Stainless Steel Tank | |
40 kHz | |
2.5 gal. | |
29 x 24 x 15 cm (11.5 x 9.5 x 6 in.) | |
M5800-E | |
6.4 kg | |
CEE 7/7 (Continental European) |