missing translation for 'onlineSavingsMsg'
Learn More
Learn More
m-Xylene, 99+%, extra pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mL
1 L
10 L
Packaging:
Glass Bottle
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | -48.0°C |
CAS Min % | 99.0 |
Color | Colorless |
CAS Max % | 100.0 |
Density | 0.8640g/mL |
Boiling Point | 139.0°C |
Quantity | 250 mL |
Flash Point | 25°C |
Infrared Spectrum | Authentic |
Packaging | Glass Bottle |
Show More |
Safety and Handling
Product Identifier
- m-Xylene
- Warning
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'