Learn More
m-Xylylenediamine, 99%, Thermo Scientific Chemicals
26.43 € - 126.63 €
Chemical Identifiers
CAS | 1477-55-0 |
---|---|
Molecular Formula | C8H12N2 |
Molecular Weight (g/mol) | 136.2 |
MDL Number | MFCD00008119 |
InChI Key | FDLQZKYLHJJBHD-UHFFFAOYSA-N |
Synonym | m-xylylenediamine, 1,3-benzenedimethanamine, 1,3-bis aminomethyl benzene, 1,3-xylylenediamine, 1,3-phenylenedimethanamine, m-xylylendiamin, 1,3-xylenediamine, mxda, m-phenylenebis methylamine, methylamine, m-phenylenebis |
PubChem CID | 15133 |
IUPAC Name | [3-(aminomethyl)phenyl]methanamine |
SMILES | C1=CC(=CC(=C1)CN)CN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10285140
|
Thermo Scientific Acros
141020050 |
5 mL | Glass bottle |
26.43 €
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10627112
|
Thermo Scientific Acros
141021000 |
100 mL | Glass bottle |
38.39 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10741925
|
Thermo Scientific Acros
141025000 |
500 mL | Glass bottle |
126.63 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1477-55-0 | |
136.2 | |
FDLQZKYLHJJBHD-UHFFFAOYSA-N | |
15133 | |
C1=CC(=CC(=C1)CN)CN |
C8H12N2 | |
MFCD00008119 | |
m-xylylenediamine, 1,3-benzenedimethanamine, 1,3-bis aminomethyl benzene, 1,3-xylylenediamine, 1,3-phenylenedimethanamine, m-xylylendiamin, 1,3-xylenediamine, mxda, m-phenylenebis methylamine, methylamine, m-phenylenebis | |
[3-(aminomethyl)phenyl]methanamine |
Specifications
1477-55-0 | |
100.0 | |
1.0300g/mL | |
>112°C | |
99% | |
C6H4(CH2NH2)2 | |
MFCD00008119 | |
1.03 | |
FDLQZKYLHJJBHD-UHFFFAOYSA-N | |
[3-(aminomethyl)phenyl]methanamine | |
15133 | |
99% | |
m-Xylylenediamine |
98.5 | |
Yellow | |
265°C (745.0 mmHg) | |
Authentic | |
C8H12N2 | |
1.5700 to 1.572 | |
13, IV, 293 | |
m-xylylenediamine, 1,3-benzenedimethanamine, 1,3-bis aminomethyl benzene, 1,3-xylylenediamine, 1,3-phenylenedimethanamine, m-xylylendiamin, 1,3-xylenediamine, mxda, m-phenylenebis methylamine, methylamine, m-phenylenebis | |
C1=CC(=CC(=C1)CN)CN | |
136.2 | |
136.2 | |
Liquid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Harmful to aquatic life with long lasting effects.<br
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 216-032-5
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.