Learn More
Applied Biosystems™ M13 Sequencing Forward Primer, HPLC
M13 Sequencing Forward Primer, HPLC
Brand:  Applied Biosystems™ A15637
Product Code. 15554374
  • 63.50 € / Each
Explore more special offers
Description
Description
Use our tool to find and order pre-designed primers for PCR and Sanger sequencing. Free of SNPs and dimers, target-specific, and suitable for universal PCR conditions, these primers can be ordered unmodified, M13-tailed, HPLC-purified, or desalted. All primers are checked by mass spectrometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Single tube, dried-down
Mass spectrometry
M13 (-21) Forward Sequencing Primer
M13 (-21) Forward Sequencing
HPLC
Solid
M13 (-21) Forward Sequencing Primer, 50nmole, HPLC Purification, 900pmole Minimal Spec Yield, 5' TGTAAAACGACGGCCAGT Sequence, Delivered in a Single tube, Dried-down, Used for BigDye™ Cycle Sequencing Primers
5' TGTAAAACGACGGCCAGT
50 nmol
BigDye™ cycle sequencing primers