Learn More
Brady™ M210 Label Printer - Carton Pack
The M210 Label Printer is an all-in-one labeler for voice and datacom, electrical and industrial use, creating clear labels that stay stuck for years- despite extreme temperatures or highly textured surfaces.
Brand:  Brady™ 152260
Product Code. 17653215
  • 120.00 € / Each
Explore more special offers
Description
Description
Brady™ M210 Label Printers provides engineered, long lasting material quality with a two-year warranty. Extremely rugged and durable, the M210 Label Printer has been drop tested and is crafted with molded rubber bumpers to provide optimal protection from falls. This printer ships in a carton pack with a Quick Start Guide and an M21-750-499 label cartridge.
  • Prints true-sized 6 to 19 mm wide identification to maximize amount of data
  • Adjust length of labels using high-performance, continuous material
  • Seven different industry specified materials that last
  • Ergonomic cutter and label grabber that holds label after cutting
  • Fast label creation with ABC keypad, graphics library, hot keys and menu functions
  • Multi-functional accessory with magnet, flashlight and retractable printer stand for easy handling
  • Smart Cell Technology for automatic label setup and formatting for wire wraps, terminal blocks, patch panels, cable flags and general labels
  • Languages: Croatian, Czech, Danish, Dutch, English Estonian, Finnish, French, German, Hungarian, Italian, Norwegian, Polish, Portuguese, Romanian, Slovakian, Slovenian, Spanish, Swedish, Turkish
Specifications
Specifications
Label Printer
Monocolor LCD
Thermal Transfer
114 mm
241 mm
64 mm
Monochrome
Croatian, Czech, Danish, Dutch, English, Estonian, Finnish, French, German, Hungarian, Italian, Norwegian, Polish, Portuguese, Romanian, Slovakian, Slovenian, Spanish, Swedish, Turkish
M210
203 dpi
2011/65/EU and 2015/863
2 Years
110/240 V
Yellow, Black
Brady Labels
4.5 in.
9.5 in.
2.5 in.
2.5 x 4.5 x 9.5 in.
Printer, Quick Start Guide and M21-750-499 Label Cartridge
16 MB
10 mm/s
0.75 kg
Data and Telecommunications Labeling, Electrical Labeling, General Identification, Warehouse Marking
AC Adaptor, Batteries, Rechargeable Battery
ABC