Learn More
Brady™ M710 Label Printer Wi-Fi, Bluetooth and BWS PWID Suite
More speed, less mess. It’s Brady™'s fastest, most advanced portable printer yet with a print speed of 76.2 mm/sec. Printer includes Wi-Fi, Bluetooth, and Brady™ WorkStation Product & Wire Identification Suite.
Brand:  Brady™ 317824
 View more versions of this product
Product Code. 17815255
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Auto-cut labels and work all day on one charge
  • Use one device for 100's of applications, printing on the toughest materials around
  • Stay connected wherever your work happens with enhanced Bluetooth and Wi-Fi
  • Get lightning-fast print speeds, plus cut labels automatically
  • Prints 4,500 labels on a full battery charge
  • Enjoy more durability with a printer that withstands 1.2 m drops, blowing sand and dust
  • Get more work done using a keypad or software to print labels
  • Print what you need, how you need – text, barcodes, shapes and images using a keypad or software
  • Get shop-to-field flexibility with label saving, sharing and data imports
  • Comes with Brady™ WorkStation Product & Wire Identification Suite, enabling you to immediately start designing professional labels
Specifications
Specifications
Printer
Color LCD Touchscreen
Plastic
4.6 in.
116.75 mm
13.59 in.
Monocolor
Printer, Hard Case, USB Cable, Quick Start Guide, Drivers, Li-ion Rechargeable Battery, AC Adapter, Users Guide, Brady WorkStation Product and Wire ID Software Suite, Media Adapter, Roll of M7C-1000-595-WT and M7-R6000 Ribbon
200 MB (Stores Up to 100,000 Labels)
76.2 mm/sec.
Bluetooth, Wi-Fi, USB 2.0, USB-C
2.45 kg
Data and Telecommunications Labeling, Laboratory Labeling, Wire and Cable Labeling
AC Adaptor, Rechargeable Battery
QWERTY (UK)
Blue
Brady Labels
Thermal Transfer
9.45 in.
240.03 mm
345.2 mm
53.77 x 71.28 mm
English (US), French, Spanish, Portuguese, German, Dutch, Norwegian, Finnish, Swedish, Italian, Danish, Bulgarian, Russian, Croatian, Hungarian, Japanese, Korean, Polish, Romanian, Slovak, Turkish, Estonian, Chinese (Traditional), Chinese (Simplified)
M710
300 dpi
5.4 lb.
FCC Part 15 Class A
2 Years
110/240 V
M710-WB-QY-UK-PWID