Learn More
Thermo Scientific™ MA(PEG)4 Methyl-PEG-Amine Compound
MA(PEG)n: methyl- and amine-capped polyethylene glycol compounds containing 4 to 24 PEG units; use for surface-modification and molecule-pegylation applications.
Brand:  Thermo Scientific™ 26111
 View more versions of this product
Product Code. 10627344
  • 988.00 € / 1g
Estimated Shipment: 06-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Thermo Scientific™ Pierce MA(PEG)n Compounds are methyl- and amine-terminated polyethylene glycol reagents, where n equals 4 to 24 PEG units, that are useful for a variety of surface-modification and molecule-pegylation applications.
MA(PEG)n compounds have the form Methyl-PEGn-Amine and are ideal for derivatizing surfaces and molecules with discrete numbers (n = 4, 8, 12, 24) of ethylene glycol groups. Use any amine-reactive chemistry (e.g., EDC or NHS ester) to PEGylate surfaces, proteins, or other molecules with a precise number of hydrophilic, nonimmunogenic PEG units.
Highlights:
  • Pure compounds of specific size ensure complete control of PEGylation
  • Straight-chain compounds with simple terminal primary amine
  • Adaptable to a variety of modification procedures
Recommended for:
  • PEGylate carboxylate or amine surfaces Add inert mass to proteins, immunogens, drug compounds and probes Improve solubility (decrease aggregation) of proteins or peptides without affecting function Protect proteins from proteolysis
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ketone
Upon receipt store product at -20°C protected from moisture. Product is shipped at ambient temperature.
Amine, Primary Amine
Liquid
Water
Pierce MA(PEG)4
Polyethylene Glycol (PEG), Blocking Groups
4 ethylene glycol units
1 g
Amine Compound