missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MabCaptureC™ MiniChrom™ Pre-packed Column

Thermo Scientific™ MabCaptureC™ MiniChrom™ columns are designed for bench-scale resin screening, process development, and sample preparation.

Brand:  Thermo Scientific™ 5943662001

 View more versions of this product

Product Code. 30019374

  • 507.00 € / 1mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

MabCaptureC™ MiniChrom™ columns are designed for bench-scale resin screening, process development, and sample preparation.

These pre-packed columns are ready to connect and have the following features:

  • Packed with MabCaptureC™ Affinity Matrix, offering high dynamic binding capacity and high resolution over a wide range of flow rates
  • Direct connection to standard chromatography systems

The 1 mL columns can be used for quick screenings of application feasibility and lab scale purification on a convenient and easy-to-use pre-packed column. The 10-cm bed height of the 5 mL column allows initial process development at a bench scale.

The pre-packed MabCaptureC™ Affinity Matrix contains engineered rProtein A that binds at the CH2-CH3 interface of the Fc-region of IgGs and shows cross binding to the VH3 domain of IgGs.

Features of MabCaptureC™ Affinity Matrix include:

  • High dynamic mAb binding capacity of > 50 g/L at 4.8 min residence time
  • Purification of mAbs and a subpopulation of scFv's and Fab fragments containing VH3 binding domains
  • Clean and sanitize with standard sodium hydroxide agents for chromatography products
  • Excellent scalability
  • Non-animal derived

Main resin characteristics:

  • Matrix: agarose-based, epoxide activated
  • Average particle size: 75 ± 10 μm
  • Ligand: recombinant C domain of Protein A
  • Ligand coupling method: epoxide coupling
  • Binding capacity: >55 g IgG/L resin at 5 min residence time and 10% breakthrough; >75 g IgG/L resin at 10 min. residence time and 10% breakthrough
  • Elution conditions: 0.1 M glycine pH 3.0, or 20–100 mM sodium citrate or acetate, pH 3.0
  • Cleaning conditions: 0.2 M sodium hydroxide for 15 minutes every cycle
  • Column dimensions: 0.8 x 2 cm (1 mL), 0.8 x 10 cm (5 mL)
  • Operating pressure: ≤ 3 bar
  • Formulation buffer: 20% (v/v) ethanol
Specifications

Specifications

1 mL
0.8 cm
2 cm
Agarose-based, Epoxide-activated
Agarose bead
1 mL
>55 g IgG/L resin (5 min. residence time and 10% breakthrough), >75 g IgG/L resin (10 min. residence time and 10% breakthrough)
75 ±10 μm
Engineered rProtein A
≤3 bar
IgG mAbs, and a subpopulation of scFv and Fab containing VH3 domain
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.