missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MAbPac™ HIC-10 HPLC Column

Separate monoclonal antibodies (mAbs) and related biologics with high resolution, excellent biocompatibility, and selectivity using the unique chemistry of this hydrophobic interaction column.

Brand:  Thermo Scientific™ 088482

237.36 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15300421

  • 238.55 € / Pack of 2
Estimated Shipment: 24-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Designed for separation of monoclonal antibodies (mAbs) and related biologics by hydrophobic interaction, the Thermo Scientific™ MAbPac™ HIC-10 LC column is a high resolution silica column. The MAbPac HIC-10 column's unique chemistry provides high resolution, excellent biocompatibility, and selectivity that is different from other HIC columns on the market.

Unique Chemistry Designed for Optimal Selectivity of mAbs

The MAbPac HIC-10 column is based on high-purity, spherical, wide-pore (1,000Å), 5μm silica particles functionalized with proprietary alkyl amide groups. Its advanced surface bonding technology leads to excellent chemical stability, high efficiency, and unique selectivity optimized for mAbs and different from other HIC phases.
  • Excellent biocompatibility and extremely low carryover
  • High resolution and high efficiency
  • Compatible with both organic solvent and aqueous mobile phases
  • Rugged column stability
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Guard Column
10 mm
5 μm
1000 Å
60°C
MAbPac
Hydrophobic Interaction
Bio-LC
4.6 mm
6000 psi (415 bar)
2 to 8
Each
Silica
Spherical, Fully Porous, Ultrapure Silica
Hydrophobic Interaction
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.