missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MAbPac™ HIC-20 HPLC Columns

Use this column to separate monoclonal antibodies (mAbs), mAb fragments and oxidized variants by hydrophobic interaction chromatography (HIC).

Brand:  Thermo Scientific™ 088555

238.94 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15334695

  • 240.13 € / Pack of 2
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Use the Thermo Scientific™ MAbPac™ HIC-20 HPLC column for separation of monoclonal antibodies (mAbs), mAb fragments and oxidized variants by hydrophobic interaction chromatography (HIC). The unique chemistry of this silica column offers high resolution and excellent selectivity. The advanced surface bonding technology provides excellent chemical stability, high resolution, and unique selectivity optimized for mAbs and different from other HIC phases.

Characteristics of the MAbPac HIC-20 Column 
This column is based on high-purity, spherical, wide-pore (1,000 Å), 5μm silica particles functionalized 
with proprietary alkyl amide groups offering: 
  • Optimal selectivity for mAbs and related variants
  • Excellent separation of mAb fragments and oxidized variants
  • High resolution and high efficiency 
  • Compatible with both organic solvent and aqueous mobile phase 
  • Rugged column stability
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Guard Column
10 mm
5 μm
1000 Å
60°C
MAbPac
Hydrophobic Interaction
Bio-LC
4.6 mm
6000 psi (415 bar)
2 to 9
Each
Silica
Spherical, High Purity Silica
Hydrophobic Interaction
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.