Learn More
Thermo Scientific™ MAbPac™ HIC-Butyl HPLC Columns
Separate monoclonal antibodies (mAbs) and antibody-drug conjugates (ADCs) with excellent biocompatibility, low carryover and high resolution.
Brand: Thermo Scientific™ 088558
1278.53 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
Description
Use the Thermo Scientific™ MAbPac™ HIC-Butyl HPLC columns to separate monoclonal antibodies (mAbs) and antibody-drug conjugates (ADCs). This column is based on hydrophilic, nonporous, 5μm polymer particles functionalized with butyl groups. The column's hydrophilic resin provides excellent biocompatibility and low carryover, while the optimal density of the butyl groups provides high resolution separation.
Features of MabPac HIC-Butyl Columns- Optimal selectivity for ADCs
- High resolution and high efficiency
- Stable at wide range of pH (2−12)
- Low carryover
- Rugged column packing
Specifications
Analytical Column | |
100 mm | |
5 μm | |
Nonporous | |
60°C | |
MAbPac | |
Hydrophobic Interaction | |
Bio-LC |
4.6 mm | |
4000 psi (275 bar) | |
2 to 12 | |
Each | |
Silica | |
Hydrophilic Polymer | |
Hydrophobic Interaction |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.