missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MAbPac™ HIC-Butyl HPLC Columns

Separate monoclonal antibodies (mAbs) and antibody-drug conjugates (ADCs) with excellent biocompatibility, low carryover and high resolution.

Brand:  Thermo Scientific™ 088559

237.36 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15354695

  • 238.55 € / Pack of 2
Estimated Shipment: 24-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Use the Thermo Scientific™ MAbPac™ HIC-Butyl HPLC columns to separate monoclonal antibodies (mAbs) and antibody-drug conjugates (ADCs). This column is based on hydrophilic, nonporous, 5μm polymer particles functionalized with butyl groups. The column's hydrophilic resin provides excellent biocompatibility and low carryover, while the optimal density of the butyl groups provides high resolution separation.

Features of MabPac HIC-Butyl Columns
  • Optimal selectivity for ADCs
  • High resolution and high efficiency
  • Stable at wide range of pH (2−12)
  • Low carryover
  • Rugged column packing
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Guard Column
10 mm
5 μm
Nonporous
60°C
MAbPac
Hydrophobic Interaction
Bio-LC
4.6 mm
4000 psi (275 bar)
2 to 12
Each
Silica
Hydrophilic Polymer
Hydrophobic Interaction
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.