Learn More
Thermo Scientific™ MAbPac™ Protein A Antibody Analysis and Purification HPLC Column, 12 μm, 4 mm x 35 mm
Column Format: Analytical Column; Pore Size: Nonporous; Column Phase: Affinity Separation
Brand: Thermo Scientific™ 082539
1200.75 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
Description
Run fast, accurate titer analysis of monoclonal antibodies (mAbs) in harvest cell cultures with the Thermo Scientific™ MAbPac™ Protein A LC Columns. This column provides high throughput and accurate analyses through a combination of low backpressure and high efficiency. Its unique format makes automation of loading, binding, elution and collection using biocompatible systems quick and easy. A nonporous, polymeric resin with a hydrophobic divinylbenzene core and a hydrophilic surface optimized for affinity separation makes this column rugged and reproducible.
Fast, Accurate mAb Titer Analysis
- Complete cycle time, including equilibration, of two minutes
- Linear over a wide concentration range (0.025 to 5mg/mL) without carryover
- Low column pressure of less than 250 psi at 2mL/min
- Excellent sample recovery
Use the MAbPac Protein A LC column for both titer analysis or concentrating and purifying IgG material for later experiments or two-dimensional (2D) chromatography. It is available in an HPLC bio-inert, 4×35mm PEEK column body.
Specifications
Analytical Column | |
35 mm | |
Spherical polymer | |
2.5 to 7.5 | |
Each | |
Protein A | |
Spherical, Nonporous Polymer | |
Affinity Separation | |
Bio-LC |
4 mm | |
1000 psi (70 bar) | |
12 μm | |
Nonporous | |
30°C | |
MAbPac | |
Antibody Analysis and Purification | |
Analytical Column |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.