missing translation for 'onlineSavingsMsg'
Learn More

Cytiva MabSelect™ Chromatography Media

Protein A affinity medium designed for capturing MAbs from large sample volumes

Brand:  Cytiva 17-5199-02


 View more versions of this product

Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11359981

  • 4367.00 € / 200mL
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • High-flow agarose matrix allows for high flow velocities at production scale
  • BioProcess medium supported for industrial applications and well-established in approved processes
  • Low non-specific binding resulting in low levels of impurities in the product eluate pool
  • Oriented coupling of ligand and optimized matrix gives high dynamic binding capacity
  • Based on highly cross-linked agarose matrix with recombinant protein A ligand
  • Ligand is engineered to favor oriented coupling and expressed in E. coli and is free of components of mammalian origin
  • Hydrophilic nature of the base matrix ensures low levels of non-specific

  • Average particle size: 85μm (d50V)
  • pH Stability cleaning-in-place (CIP): 2 to 12
  • pH Stability working range: 3 to 10
  • Ligand: recombinant protein A (E.coli)
  • Matrix: highly cross-linked agarose
  • Binding capacity/mL chromatography medium: ∽30mg human lgG/mL at 2.4 min. residence time
  • Pressure/Flow specification: <500cm/hr., <200 kPa, BPG 300, bed height 20cm

TRUSTED_SUSTAINABILITY
Specifications

Specifications

200 mL
2°C to 8°C
MabSelect Media
85 μm
20 cm
<200 kPa
3 to 10
For Industrial AppliCatIons and Well-Established in Approved Processes
30 mg Human lgG/mL (at 2.4 min. residence time)
<500 cm/Hoμr
7.87 in.
Certificate of Analysis
Highly Cross-Linked Agarose
Recombinant Protein A (E. coli)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • MabSelect, 200 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers