missing translation for 'onlineSavingsMsg'
Learn More

Cytiva MAbTrap™ Kit

For small scale affinity purification of IgG from ascites fluid, serum, and cell culture supernatants in less than 15 min. using a syringe

Brand:  Cytiva 17-1128-01


Product Code. 10686505

  • 518.00 € / 1 set
In Stock
to see stock.
Explore more special offers
Add to basket

Includes: One HiTrap Protein G HP 1mL column with 50mL 10X binding buffer, 15mL 10X elution buffer, 25mL neutralizing buffer, connectors, and syringe
This item is not returnable. View return policy

Description

Description

MAbTrap Kit contains everything necessary to perform a complete one step purification without using complicated equipment, eliminating time-consuming buffer preparation and column packing. The kit includes one HiTrap column, prepacked with Protein G Sepharose High Performance with sufficient buffer concentrate for 20 purifications.

  • MAbTrap Kit contains everything necessary to perform a complete one-step purification without using complex equipment
  • Convenient kit for purification of IgG from monoclonal and polyclonal material using a syringe in less than 15 min.
  • Kit contains one HiTrap Protein G HP column prepacked with Protein G Sepharose High Performance and enough buffer concentrate for 20 preparations with a syringe
  • Recombinant protein G lacking albumin binding region

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 mL
MAbTrap is an affinity chromatography kit, with everything required for small-scale IgG purification from ascites fluid, serum, and cell culture supernatants.
4 to 8°C, 20% Ethanol
For purification of IgG from monoclonal and polyclonal material using a syringe in <15 min.
>25 mg/mL Human IgG Medium
4 mL/min.
1 Kit
MAbTrap Kit
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P235-Keep cool.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol, trometamol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers