Learn More
Thermo Scientific™ MacConkey Agar No. 3
Detect and enumerate coliform organisms, such as E. coli, while isolating and detecting Salmonella and Shigella spp. in pathological and food specimens.
27.65 €
Specifications
Format | 90mm Monoplate |
---|---|
Media Type | MacConkey Agar No. 3 |
Product Type | Agar |
Packaging Type | Flow Wrapped - Single Layer |
Quantity | 10/Pk. |
Product Code | Brand | Quantity | Format | Media Type | Manufacturing Origin | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Format | Media Type | Manufacturing Origin | Price | Quantity & Availability | ||
12992618
|
Thermo Scientific™
PO0495A |
10/Pk. | 90mm Monoplate | MacConkey Agar No. 3 | United Kingdom |
27.65 €
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Detect and enumerate coliform organisms, including E. coli, while isolating and detecting Salmonella and Shigella spp. in both pathological and food specimens using Thermo Scientific™ MacConkey Agar No. 3. One of the most famous formulations for detection of coliforms Salmonella and Shigella has improved its selectivity to Gram-positive organisms and is also a good medium for stool diagnostics.
Due to the inclusion of a specially prepared fraction of bile salts in addition to crystal violet, MacConkey Agar No. 3 is a highly selective medium for differentiating between coliforms and lactose and non-lactose fermenters while gram-positive cocci are completely inhibited.Urinary tract infections, bacteremia, meningitis, and diarrheal disease are the most frequent clinical syndromes associated with Escherichia coli. Escherichia coli O157:H7 is associated with hemorrhagic colitis and hemolytic-uremic syndrome (HUS). Escherichia coli is an aerobic, gram-negative bacillus most frequently isolated from the intestines of humans and animals. As a result, it is used as an indicator of fecal contamination in water sources. This pathogen readily grows on a variety of culture media.
Salmonella and Shigella are major causes of bacterial enteric illness. Transmission occurs via the fecal-oral route, person-to-person contact, or by the ingestion of contaminated food or water. A relatively small number of organisms ingested is capable of causing symptomatic infection. Salmonella and Shigella are aerobic, gram-negative, oxidase-negative bacilli. Some Salmonella and all Shigella are nonmotile. These organisms grow on standard media, such as MacConkey Agar, SS Agar, and XLD Agar. Enrichment broths should also be used when culturing for these organisms to allow for maximum recovery.
This product may not be available in all countries or territories. Please consult your local representative for availability.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
90mm Monoplate | |
Agar | |
10/Pk. |
MacConkey Agar No. 3 | |
Flow Wrapped - Single Layer |
Safety and Handling
missing translation for 'shelfLife' : 14 Weeks
Your input is important to us. Please complete this form to provide feedback related to the content on this product.