missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Macrosep™ Advance Centrifugal Devices with Omega Membrane
Macrosep Advance Centrifugal Devices with Omega Membrane quickly concentrates up to 20 mL of biological sample.
Brand: Cytiva MAP100C38
1382.25 EUR valid until 2025-12-31
Use promo code "24320" to get your promotional price.
This item is not returnable.
View return policy
Description
- Exchange buffer or remove salt of chromatography eluates and gradient fractions
- For research applications only
Specifications
Clear | |
PES | |
Non-sterile | |
5 to 20 mL (Sample) |
EFA: 7.2 cm2 | |
100 kDa | |
100/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction