missing translation for 'onlineSavingsMsg'
Learn More

MAG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14386-1-AP-150UL

 View more versions of this product

Product Code. 16863004

  • 410.00 € / 150µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The protein encoded by this gene is a type I membrane protein and member of the immunoglobulin superfamily. It is thought to be involved in the process of myelination. It is a lectin that binds to sialylated glycoconjugates and mediates certain myelin-neuron cell-cell interactions. Two alternatively spliced transcripts encoding different isoforms have been described for this gene.
Specifications

Specifications

MAG
Polyclonal
Unconjugated
MAG
GMA, MAG, myelin associated glycoprotein, S MAG, SIGLEC 4A, SIGLEC4A
Rabbit
Antigen Affinity Chromatography
RUO
17136, 29409, 4099
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P07722, P20916, P20917
Mag
MAG Fusion Protein Ag5845
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers