Learn More
R&D Systems™ MagCellect Plus Human CD44+ Cell Isolation Kit
Functions as a co-receptor that modifies activity of receptors including Met and the ErbB family.
Brand:  R&D Systems™ MAGH122
Product Code. 18796088
  • 891.00 € / Each
Explore more special offers
Description
Description
CD44 is a ubiquitously expressed receptor for hyaluronan and exerts control over cell growth, migration, and tumor progression. With variable N and O-glycosylation and splicing within the stalk, CD44 can range from 80 to 200 kDa. CD44 binds hyaluronan and other ligands within the extracellular matrix and can function as a “platform” for growth factors and metalloproteinases. It can function as a co-receptor that modifies activity of receptors including Met and the ErbB family tyrosine kinases. The intracellular domain of CD44 links the plasma membrane to the actin cytoskeleton via the ERM proteins, Ezrin, Radixin and Moesin.
Specifications
Specifications
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
ELISA
Cell Isolation Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only. Not for use in Humans.