Learn More
R&D Systems™ MagCellect Plus Human CD45+ Cell Isolation Kit
The MagCellect™ Plus Human CD45+ Cell Isolation Kit is designed to isolate CD45 expressing cells via a positive selection principle. The resulting cell preparation is highly enriched with CD45+ cells. Typical purity of recovered CD45+ cells ranges from 80-95%.
Brand:  R&D Systems™ MAGH125
Product Code. 30002118
  • 857.00 € / 1 set
Explore more special offers
Description
Description
A mononuclear cell suspension is first incubated with the Human CD45 Biotinylated Antibody that targets the desired cells. MagCellect™ Streptavidin (SA) Ferrofluid is next added to the reaction that allows the SA coated nanoparticles to interact with the cells tagged with the monoclonal antibodies. The tube containing the cell suspension is then placed within a magnetic field. Magnetically tagged cells will migrate toward the magnet (desired cell fraction), leaving the untagged cells (unwanted cell population) in suspension to be harvested by aspiration while the tube remains in the magnetic field. The tube containing the magnetically selected (desired) cells is then removed from the magnet and the cells are resuspended in MagCellect™ Plus Buffer or tissue culture media. The enriched cell preparation is then available for a variety of applications including tissue culture, immune status monitoring and flow cytometry.
Kit Contents:
  • Human CD45 Biotinylated Antibody (625 μL of Human CD45 biotinylated antibody in a phosphate buffered solution containing BSA and preservative)
  • Human CD45 Alexa Fluor™ 647 Antibody (125 μL of Alexa Fluor™ 647-conjugated mouse anti-human CD45 Detection antibody)
  • Streptavidin Ferrofluid (1.25 mL of a solution containing BSA and preservatives)
  • 2 bottles (50 mL/bottle) of a 10X concentrated buffer
Specifications
Specifications
B220, CD45, CD45 antigen, CD45R, EC 3.1.3.48, LCA, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, PTPRC, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
1 Kit
5788