missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Scienceware™ Magic Touch Ice Buckets

Unbreakable under normal use conditions. Bel Art™ Magic Touch™ Ice Buckets have excellent insulating properties for keeping temperature sensitive samples in an optimal environment.

Brand:  Bel-Art™ Scienceware™ M18848-2092

 View more versions of this product

Product Code. 12328687

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Temperature range: -196° to 100°C (-321° to 212°F)
  • Use with ice, dry ice, dry ice alcohol slurries or liquid nitrogen
  • Non-sweating
  • Impervious to moisture and odors
  • Expanded Polymer Foam is chemical resistant
  • Unbreakable under normal use conditions
  • Flared edge for easy carrying
  • Lid can be cut to make openings for thermometers, cooling coils, flasks etc. 
  • Available in 2 sizes
  • Round floating racks available for 1.5ml and 15ml tubes
Specifications

Specifications

The use of ice, dry ice, dry ice alcohol slurries or liquid nitrogen.
Ice Bucket
2.5 L
Polymer
Black
-196°C to +90°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers