missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady Magicard™ 300/600-Cleaning Roller Kit
Click to view available options
Unit Size:
Pack of 5
Description
- Dust-Free printing.
- Prepares the card for high quality printing.
- Lasts for a complete roll or dye film.
Specifications
Specifications
| Product Type | Cleaning Kit |
| For Use With (Equipment) | Magicard 300, Magicard 600 |
| Description | Cleaning Roller Kit for Magicard 300/600 Printer |
| Format | Cleaning Kit |
| Includes | 5 Cleaning Rollers, 1 Reusable Roller Bar |
| For Use With (Application) | Healthcare and Hospital labelling;General identification;Facility identification;Security identification |
| Quantity | 5 Pk. |
| Alternate Part Number | 3633-0054 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction