Learn More
Applied Biosystems™ MagMAX CORE Nucleic Acid Purification Kit + Mechanical Lysis Module
Shop All Applied Biosystems ProductsDescription
The Applied Biosystems™ MagMAX™ CORE sample preparation solution is a universal sample preparation system designed to meet current and future veterinary testing needs. This modular technology is optimized for the purification of nucleic acids from a wide range of sample matrices commonly submitted for veterinary diagnostic testing. The MagMAX™ CORE Mechanical Lysis Module is offered as a supplement to the MagMAX™ CORE Nucleic Acid Purification Kit to aid in the processing of hard-to-lyse organisms like Mycobacterium avium paratuberculosis (MAP) and bovine tuberculosis in feces, milk, and environmental samples.
Features include:
- Convenience—a complete solution using the same CORE chemistry
- Confidence—higher-quality nucleic acids from difficult-to-lyse samples, including both small (0.3 g) and large (2 to 5 g) volume feces samples
- Flexible—purchase only what you need to lyse MAP and other hard-to-lyse organisms
The MagMAX™ CORE Nucleic Acid Purification Kit and the MagMAX™ CORE Mechanical Lysis Module help make your lab more efficient through greater convenience, consistent performance, and time and cost savings.
Specifications
Specifications
| Content And Storage | • 1 x 50 mL MagMAX CORE Lysis Solution • 1 x 45 mL MagMAX CORE Binding Solution • 1 x 60 mL MagMAX CORE Wash Solution 1 • 1 x 60 mL MagMAX CORE Wash Solution 2 • 1 x 12 mL MagMAX CORE Elution Buffer • 1 x 2.2 mL MagMAX CORE Magnetic Beads • 1 x 1.25 mL MagMAX CORE Proteinase K • 1 x 45 mL MagMAX CORE Clarifying Solution • 100 MagMAX CORE Bead Beating Tubes (2 mL) |
| Sample Type | Bacteria, Blood, Buccal Samples, Cells, DNA, RNA, Tissue, Virus |
| For Use With (Application) | Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR |
| High-throughput Compatibility | Automated Protocols, Manual Protocols, Multiplexing |
| Product Line | MagMAX |
| Quantity | 100 Tests |
| Shipping Condition | Room Temperature |
| Target | Total RNA, Pathogen DNA⁄RNA, Genomic DNA, Viral DNA⁄RNA |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.