Learn More
Applied Biosystems™ MagMAX™ Microbiome Lysis Solution
This lysis buffer is included in the MagMAX Microbiome Ultra Nucleic Acid Isolation Kit.
Brand:  Applied Biosystems™ A42361
459.00 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Product Code. 16356582
  • 612.00 € / 1000 reactions
Explore more special offers
Description
Description
This lysis buffer is included in the MagMAX Microbiome Ultra Nucleic Acid Isolation Kit (Cat. Nos. A42357, and A42358). It is made available separately for applications that require more lysis solution than is provided in the kit.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store at room temperature.
Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
MagMAX™
Room Temperature
Bacteria, Blood, Buccal Samples, Cells, Fungi, Liquid Samples (e.g. Serum), Viral Samples, Yeast
High-throughput Compatible, Manual Protocols, Automated Protocols
800 mL
Viral DNA⁄RNA, Pathogen DNA⁄RNA, Total Nucleic Acids