missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ Plant RNA Isolation Kit

The MagMAX Plant RNA Isolation Kit is designed for automated high-throughput or manual purification of total RNA from wide variety of plant species and tissue types.

333.00 € - 1718.00 €

Specifications

Sample Type Plant Samples
Shipping Condition Room Temperature
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15786048
View Documents
Applied Biosystems™
A33784
96 Preps
333.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
15796048
View Documents
Applied Biosystems™
A33899
384 Preps
1286.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
17153621
View Documents
Applied Biosystems™
A47157
1000 Preps
1718.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The MagMAX Plant RNA Isolation Kit is designed for automated high-throughput or manual purification of total RNA from wide variety of plant species and tissue types. It utilizes paramagnetic bead technology, enabling high yields and robust performance. The high binding capacity, uniform particle size, and rapid magnetic response of MagMAX magnetic beads makes the technology ideal for high-throughput automatic nucleic acid purification, as well as for manual nucleic acid purification by low- throughput users. The resulting high quality purified RNA is free of proteins, nucleases, and other contaminants or inhibitors, and can be used in a wide range of downstream applications, such as RT-PCR, RT-qPCR, and other enzymatic reactions.

Versatile—suitable for any plant type and part, including woody, polyphenol-rich, and lignified samples
High yields of pure total RNA
High quality—isolates intact total RNA, free of inhibitors and gDNA contamination. Isolated RNA is suitable for long term storage at -20°C.
Flexible—easily adjustable for high-throughput automation platforms and manual processing

Applications
The MagMAX Plant RNA Isolation Kit is designed for fast extraction of high purity RNA suitable for all conventional molecular biology applications, including:
• RT-PCR
• RT-qPCR
• Northern blotting
• Nuclease protection assay

Benefits of MagMAX magnetic bead–based purification
The magnetic bead–based purification format of the kit allows for easy scaling of the number of samples processed either manually with a magnetic stand or with a magnetic particle processor such as the KingFisher Duo Prime or KingFisher Flex processors.

Magnetic beads offer many benefits compared to other technologies for isolating nucleic acids. Beads bind the RNA and DNA more efficiently than glass fiber filters, resulting in higher and more consistent yields. Additionally, because filters and vacuum manifolds are not used, there is no risk of undigested tissue particulates clogging these items during the extraction process. This clogging issue is of particular concern with fibrous plant tissue samples that that are not fully lysed during the disruption and homogenization steps.

Optimized bead surface chemistry
The Dynabeads MyOne Carboxylic Acid utilized in the MagMAX Plant RNA Isolation Kit has an optimized surface chemistry with high specific surface area that allows efficient kinetics and high sensitivity in nucleic acid capture. These aspects make Dynabeads magnetic technology ideal for the recovery of the RNA targets found in a variety of plant samples. Once captured, the RNA is then eluted in 100 μL and ready for downstream applications.

Specifications

Specifications

Plant Samples
Room Temperature
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.