missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosytstems™ MagMAX™ Prime Viral/Pathogen NA Isolation Kit

The MagMAX Prime Viral/Pathogen NA Isolation Kit is a magnetic-bead-based nucleic acid purification kit for the isolation and purification of viral, bacterial, yeast, and fungal nucleic acid from biological specimens.

Brand:  Applied Biosytstems™ A58145

867.00 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.



Product Code. 17984661

  • 1020.00 € / 600 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The MagMAX Prime Viral/Pathogen NA Isolation Kit is a magnetic-bead-based nucleic acid purification kit for the isolation and purification of viral, bacterial, yeast, and fungal nucleic acid from biological specimens.

Features of the MagMAX Prime Viral/Pathogen NA Isolation Kit include:
• Compatible with multiple downstream assays
• Improved reagent that maximizes nucleic acid yield from routinely tested pathogens without the need for additional lysis
• Streamlined workflow accommodating various sample types and scalable sample input volume that can be run on a single plate
• Single purification protocol for various biological specimens including swabs, serum, plasma, and urine
• Designed to work with the KingFisher Flex and KingFisher Apex magnetic particle processors with the ability to purify 96 samples in under 60 minutes
• Sustainable product with biodegradable ingredients and reduced plastic usage
• Available accessories with optimized protocol and KingFisher script to extract nucleic acids from challenging organisms and stool sample type

The kit is intended for use by qualified and trained clinical laboratory personnel specifically instructed and trained in automated magnetic bead purification techniques.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Magnetic

• 160 mL Binding Solution
• 300 mL Wash I Solution
• 6 mL Binding Beads
• 3 mL Proteinase K
• 36 mL Elution Buffer
• 30 μL Proteinase K dye

Store at 15–25°C.

Beads
Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
Isolate and purify viral, bacterial, parasitic, and fungal nucleic acid
DNA/RNA
Reagent
200–400 μL
Viral, Bacterial, Parasitic, and Fungal Nucleic Acid
Clear
Kit
Swabs in Transport Media, Saliva, Urine, Serum, Plasma
High-throughput Compatible, Automated Protocols
Up to 600 Reactions
MagMAX™
Up to 600 reactions
Room Temperature
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • MagMAX Prime Viral/Pathogen NA Isolation Kit

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 C
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Proteinase, Thiocyanic acid, guanidine

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.