Learn More
Applied Biosystems™ MagMAX™ Viral/Pathogen Wash Solution
This wash solution is included in the MagMAX Viral/Pathogen Nucleic Acid Isolation kits and MagMAX Microbiome Ultra Nucleic Acid Isolation Kit.
Brand:  Applied Biosystems™ A42360
468.00 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Product Code. 16305035
  • 624.00 € / 1000 reactions
Explore more special offers
Description
Description
This wash solution is included in the MagMAX Viral/Pathogen Nucleic Acid Isolation kits and MagMAX Microbiome Ultra Nucleic Acid Isolation Kit (Cat. Nos. A42352, A42356, A42357, and A42358). It is made available separately for applications that require more wash solution than is provided in the kit.
Important: This wash solution may develop inert white or brown particulates that float in solution. This is not a cause for concern and does not negatively affect performance.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Serum, Viral Samples, Buccal Samples, Blood, Cells, Bacteria, Fungi, Yeast
Store at room temperature.
High-throughput Compatible, Manual Protocols, Automated Protocols
Viral DNA⁄RNA, Pathogen DNA⁄RNA, Total Nucleic Acids
Room Temperature
1000 mL
Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
MagMAX™
1000 mL