missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ Wastewater Ultra Nucleic Acid Isolation Kit

The MagMAX Wastewater Ultra Nucleic Acid Isolation Kit enables efficient, high-throughput nucleic acid extraction from wastewater samples.

Brand:  Applied Biosystems™ A52606

329.25 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.



Product Code. 17321909

  • 439.00 € / 100 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • The MagMAX Wastewater Ultra Nucleic Acid Isolation Kit enables efficient, high-throughput nucleic acid extraction from wastewater samples
  • The kit utilizes MagMAX magnetic-bead technology, ensuring reproducible recovery of high-quality nucleic acid compatible with a broad range of applications
  • Wastewater and sewage surveillance is a useful tool for monitoring the regional spread of viral and bacterial pathogens
  • Surveillance at different levels of operatio has been shown to be a reliable way to detect the SARS-CoV-2 virus before it becomes widespread in a population
  • Wastewater surveillance can be used to monitor other viral targets such as influenza and norovirus
  • However, wastewater and sewage samples are some of the most challenging when it comes to extraction of RNA and DNA

Features of the MagMAX Wastewater Ultra Nucleic Acid Isolation Kit include:

  • Automation-ready protocols designed for KingFisher Flex, KingFisher APEX, and KingFisher Duo systems
  • Fast procedure allows for 24–96 samples to be processed in ∼45 minutes
  • Compatible with wastewater and sewage concentrated via ultracentrifugation, precipitation, filtration, or bead-based viral enrichment and non-concentrated samples
  • Includes methods for 200 μL to 500 mL starting volumes of wastewater
  • Elution volumes ranging from 50 to 100 μL
TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 80 mL Lysis Buffer; room temperature
• 50 mL Binding Solution; room temperature
• 200 mL Wash Buffer; room temperature
• 25 mL Elution Solution; room temperature
• 4 mL Proteinase K; room temperature
• 2 mL DNA/RNA Binding Beads; room temperature

Sludge, Wastewater
RNA and DNA
RT-PCR, qPCR, cDNA library construction, NGS, microarray analysis, blot hybridization, Northern/Southern blotting, in vitro translation, nuclease protection assays, nucleic acid labeling, hybridization
100 preps
Up to 29 min. when using automated protocol
Magnetic Bead
50 to 200 μL
KingFisher™ Duo Prime, KingFisher™ Flex, KingFisher™ Apex
High-throughput Compatible
Room Temperature
Sludge: ≤200 mg
Wastewater: ≤500 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • MagMAX Wastewater Ultra Nucleic Acid Isolation Kit

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 C
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Thiocyanic acid, Proteinase

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For research use only. Not for use in diagnostic procedures.