missing translation for 'onlineSavingsMsg'
Learn More
Magnesium bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals Product Code.: 15402747

Magnesium bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals

Product Code. 15402747
5 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15402747

Brand: Thermo Scientific Alfa Aesar 046667.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Magnesium bis(trifluoromethylsulfonyl)imide is a strong Lewis acid and used as a catalyst in organic synthesis. It is an electrolyte, which shows high anodic stability used in rechargeable magnesium batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133395-16-1
Molecular Formula C4F12MgN2O8S4
Molecular Weight (g/mol) 584.576
MDL Number MFCD01321278
InChI Key DMFBPGIDUUNBRU-UHFFFAOYSA-N
Synonym bis bis trifluoromethylsulfonyl amino magnesium, magnesium bis trifluoromethanesulfonimide, magnesium ii bis trifluoromethanesulfonyl imide, magnesium ii triflimide, magnesium bistrifluoromethanesulfonimide, magnesium bis trifluoromethylsulfonyl imide, magnesium 2+ bis bistriflylimide anion, bis trifluoromethanesulfonyl imide magnesium ii saltShow MoreShow Less
PubChem CID 11851070
IUPAC Name magnesium;bis(trifluoromethylsulfonyl)azanide
SMILES C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Mg+2]

Specifications

Melting Point >200°C
Color White to Yellow
Linear Formula [(CF3SO2)2N]2Mg
Quantity 5 g
UN Number UN1759
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 584.58
Physical Form Solid
Chemical Name or Material Magnesium bis (trifluoromethylsulfonyl)imide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY16I045 Certificate TypeCertificate of Analysis Date11/01/2023 Catalog Number046667.06,046667.03,046667.MD
Lot NumberY05H068 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number046667.06,046667.03,046667.MD
Lot NumberX29G056 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number046667.06,046667.03,046667.MD
Lot NumberX27D080 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number046667.06,046667.03,046667.MD
Lot NumberN23B029 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number046667.06,046667.03,046667.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Magnesium bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.