Learn More
Magnesium bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals
Description
Magnesium bis(trifluoromethylsulfonyl)imide is a strong Lewis acid and used as a catalyst in organic synthesis. It is an electrolyte, which shows high anodic stability used in rechargeable magnesium batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | >200°C |
Color | White to Yellow |
Linear Formula | [(CF3SO2)2N]2Mg |
Quantity | 250 mg |
UN Number | UN1759 |
Sensitivity | Moisture sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 584.58 |
Physical Form | Solid |
Chemical Name or Material | Magnesium bis (trifluoromethylsulfonyl)imide |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Y16I045 | Certificate of Analysis | 11/01/2023 | 046667.06,046667.03,046667.MD |
Y05H068 | Certificate of Analysis | 12/04/2022 | 046667.06,046667.03,046667.MD |
X29G056 | Certificate of Analysis | 22/11/2021 | 046667.06,046667.03,046667.MD |
X27D080 | Certificate of Analysis | 22/11/2021 | 046667.06,046667.03,046667.MD |
N23B029 | Certificate of Analysis | 22/11/2021 | 046667.06,046667.03,046667.MD |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.