missing translation for 'onlineSavingsMsg'
Learn More

Magnesium carbonate basic, heavy, extra pure

Product Code. 10177180 Shop All Thermo Scientific Chemicals Products
missing translation for 'orderingAttributeHoverText'
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic Drum
Plastic bottle
missing translation for 'unitSize'
10kg
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10177180

missing translation for 'mfr': Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Magnesium carbonate basic, heavy, 40 to 45% (MgO), CMgO3, CAS Number-39409-82-0, 500g, loss on calcination: 55 to 60%, White, 208-915-9, 84.31, Extra Pure, MgCO3, 15,5724, MFCD00011107, Plastic bottle, Powder, OM2470000 | CAS: 39409-82-0 | C4H10Mg5O18 | 467.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39409-82-0
Molecular Formula C4H10Mg5O18
Molecular Weight (g/mol) 467.63
MDL Number MFCD00149783
InChI Key YMKSTRLFWJQKFP-UHFFFAOYSA-D
IUPAC Name pentamagnesium(2+) tetrahydrate dihydroxide tetracarbonate
SMILES O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 500 g
Linear Formula MgCO3
Merck Index 15,5724
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in ethanol, soluble in acids
Formula Weight 84.31
Percent Purity 40 to 45% (MgO)
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Magnesium carbonate basic, heavy
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.