missing translation for 'onlineSavingsMsg'
Learn More

Magnesium chloride, 1M aq. soln., Thermo Scientific Chemicals

Product Code. 15414799
Click to view available options
Quantity:
50 mL
100 mL

Magnesium chloride, 1M aq. soln., sterile is used as an excellent magnesium ion source for synthesis and biology research. Magnesium chloride is widely used to supply the magnesium ion in various molecular biology applications, including PCR reactions and buffers. It was used to prepare buffer to lyse cells for immunoprecipitation1 and to prepare serum-free medium for electrophysiological studies of rat brain slice cultures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium chloride, 1M aq. soln., sterile is used as an excellent magnesium ion source for synthesis and biology research. Magnesium chloride is widely used to supply the magnesium ion in various molecular biology applications, including PCR reactions and buffers. It was used to prepare buffer to lyse cells for immunoprecipitation1 and to prepare serum-free medium for electrophysiological studies of rat brain slice cultures.

Solubility
Soluble in water (542 mg/ml at 20°C), alcohol, methanol (15.5 g/100g at 0°C), and ethanol (3.61 g/100g at 0°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Very hygroscopic.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7786-30-3
Molecular Formula Cl2Mg
Molecular Weight (g/mol) 95.21
MDL Number MFCD00011106
InChI Key TWRXJAOTZQYOKJ-UHFFFAOYSA-L
Synonym magnesium chloride, magnesium chloride anhydrous, mgcl2, magnesium chloride, anhydrous, magnesium 2+ ion dichloride, chloromagnesite, magnesiumchlorid, magnesium ii chloride
PubChem CID 5360315
ChEBI CHEBI:6636
SMILES [Mg++].[Cl-].[Cl-]

Specifications

Physical Form Liquid
Quantity 100 mL
Linear Formula MgCl2
Solubility Information Soluble in water (542mg/ml at 20°C),alcohol,methanol (15.5g/100g at 0°C),and ethanol (3.61g/100g at 0°C).
IUPAC Name magnesium(2+) dichloride
Formula Weight 95.22
Concentration or Composition (by Analyte or Components) 1M aq. soln.
Chemical Name or Material Magnesium chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'