missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Chloride, 6-Hydrate, Crystal, U.S.P. - F.C.C., J.T. Baker™

Product Code. 15568004
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7791-18-6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Physical Form Crystals
Quantity 500 g
Molecular Formula MgCl2·6H2O
Formula Weight 203.3
Grade FCC/USP
Packaging Poly Bottle
Specific Gravity 1L = 1.57kg
Chemical Name or Material Magnesium Chloride, 6-Hydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.