missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Chloride, 6-Hydrate, Crystal, Ultrapure Bioreagent, J.T. Baker™
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Description
- For Molecular Biology Applications
Specifications
Specifications
CAS | 7791-18-6 |
Physical Form | Crystals |
Quantity | 500 g |
Molecular Formula | Cl2H12MgO6 |
MDL Number | MFCD00149781 |
InChI Key | DHRRIBDTHFBPNG-UHFFFAOYSA-L |
SMILES | O.O.O.O.O.O.[Mg++].[Cl-].[Cl-] |
IUPAC Name | magnesium(2+) hexahydrate dichloride |
Molecular Weight (g/mol) | 203.30 |
Formula Weight | 203.3 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'