missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Chloride Hexahydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

Product Code. 10647032
Click to view available options
Quantity:
500 g
1 kg
2 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Melting Point 117°C
Color White
pH 5 to 6.5
Physical Form Powder/Solid
Quantity 500 g
Formula Weight 203.31g/mol
Packaging Plastic powder jar
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'