missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Chloride Hexahydrate, Honeywell Fluka™
Puriss., meets analytical specification of BP, FCC, European Pharmacopoeia, E511, ≤0.0001% Al, 99-101%,
77.60 € - 863.02 €
Chemical Identifiers
CAS | 7791-18-6 |
---|---|
Molecular Formula | Cl2H12MgO6 |
Molecular Weight (g/mol) | 203.30 |
MDL Number | MFCD00149781 |
InChI Key | DHRRIBDTHFBPNG-UHFFFAOYSA-L |
IUPAC Name | magnesium(2+) hexahydrate dichloride |
SMILES | O.O.O.O.O.O.[Mg++].[Cl-].[Cl-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Prezzo | Quantità e disponibilità | |||||
15671260
|
Honeywell-Fluka
13152-1KG |
1 kg |
77.60 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15681260
|
Honeywell-Fluka
13152-2.5KG |
2.5 kg |
172.60 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15601270
|
Honeywell-Fluka
13152-6X1KG |
6 x 1 kg |
396.39 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15691260
|
Honeywell-Fluka
13152-6KG |
6 kg | N/A | ||||||
15611270
|
Honeywell-Fluka
13152-6X2.5KG |
6 x 2.5 kg |
863.02 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Identificatori chimici
7791-18-6 | |
203.30 | |
DHRRIBDTHFBPNG-UHFFFAOYSA-L | |
O.O.O.O.O.O.[Mg++].[Cl-].[Cl-] |
Cl2H12MgO6 | |
MFCD00149781 | |
magnesium(2+) hexahydrate dichloride |
Specifica
7791-18-6 | |
MgCl2 · 6H2O | |
NONH for all modes of transport | |
O.O.O.O.O.O.[Mg++].[Cl-].[Cl-] | |
203.30 | |
99 to 101% | |
Magnesium Chloride Hexahydrate, Meets analytical specification of Ph. Eur.BP, FCC |
Cl2H12MgO6 | |
MFCD00149781 | |
DHRRIBDTHFBPNG-UHFFFAOYSA-L | |
magnesium(2+) hexahydrate dichloride | |
203.3g/mol | |
puriss. |
Sicurezza e movimentazione
missing translation for 'einecsNumber' : 232-094-6
missing translation for 'rtecsNumber' : OM2975000