missing translation for 'onlineSavingsMsg'
Learn More

Magnesium foil, 1.0mm (0.04in) thick, hard, 99.9% (metals basis), Thermo Scientific Chemicals

212.06 € - 964.80 €

Chemical Identifiers

CAS 7439-95-4
Molecular Formula Mg
Molecular Weight (g/mol) 24.30
MDL Number MFCD00085308
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
Synonym magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
PubChem CID 5462224
ChEBI CHEBI:25107
IUPAC Name magnesium
SMILES [Mg]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11328568
View Documents
Thermo Scientific Alfa Aesar
040604.FF
25 x 25 mm
212.06 €
25mm
Please to purchase this item. Need a web account? Register with us today!
11400321
View Documents
Thermo Scientific Alfa Aesar
040604.FI
50 x 50 mm
394.97 €
Each
Please to purchase this item. Need a web account? Register with us today!
11318568
View Documents
Thermo Scientific Alfa Aesar
040604.GH
100 x 100 mm
964.80 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7439-95-4
24.30
JLVVSXFLKOJNIY-UHFFFAOYSA-N
5462224
magnesium
Mg
MFCD00085308
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
CHEBI:25107
[Mg]

Specifications

7439-95-4
MFCD00085308
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
[Mg]
24.30
CHEBI:25107
Odorless
Foil
Magnesium foil, 1.0mm (0.04 in.) thick, Hard, Temper: as rolled
Mg
UN1869
JLVVSXFLKOJNIY-UHFFFAOYSA-N
magnesium
5462224
99.9%
≈1.1g/25x25mm
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H251-H261
Flammable solid.
Self-heating: may catch fire.
In contact with water releases flammable gas.

P210-P223-P231+P232-P235+P410-P240-P241-P280-P335+P334-P370+P378s-P501c

H228-H252-H261

missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: MAGNESIUM

missing translation for 'einecsNumber' : 231-104-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only