missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium oxide, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, E 530, light, 98.0-100.5% (calc. for dried substance), Honeywell Fluka™
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
80.68 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
20 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 1 kg |
Linear Formula | MgO |
UN Number | NONH for all modes of transport |
Formula Weight | 40.3g/mol |
Percent Purity | 98 to 100.5% (calc. to the dried substance) |
Grade | puriss. |
Chemical Name or Material | Magnesium Oxide, Meets analytical specification of Ph. Eur.BP, USP, FCC |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'