missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Perchlorate, puriss p.a, ≥99.0% (calc. based on dry substance, KT), Honeywell Fluka™

Product Code. 15606080
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss., free-flowing powder,≥99.0% (calc. based on dry substance, KT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-81-8
Molecular Formula Cl2MgO8
Molecular Weight (g/mol) 223.197
MDL Number MFCD00011102
InChI Key MPCRDALPQLDDFX-UHFFFAOYSA-L
Synonym anhydrone, magnesium perchlorate, dehydrite, perchloric acid, magnesium salt, anhydrous magnesium perchlorate, unii-7n77z541yf, hsdb 661, magnesium diperchlorate, magnesium perchlorate, anhydrous, perchlorate de magnesium french
PubChem CID 24840
IUPAC Name magnesium;diperchlorate
SMILES [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Mg+2]

Specifications

Physical Form Solid
Quantity 100 g
Linear Formula Mg(ClO4)2
UN Number UN1475
Formula Weight 223.21g/mol
Percent Purity ≥99.0% (calc. based on dry substance, KT)
Packaging Poly Bottle
Chemical Name or Material Magnesium perchlorate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'