missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Perchlorate, puriss p.a, ≥99.0% (calc. based on dry substance, KT), Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Physical Form | Solid |
Quantity | 500 g |
Linear Formula | Mg(ClO4)2 |
UN Number | UN1475 |
Formula Weight | 223.21g/mol |
Percent Purity | ≥99.0% (calc. based on dry substance, KT) |
Packaging | Poly Bottle |
Chemical Name or Material | Magnesium perchlorate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'